Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCTCCAGCAGCTGCAGCAGATG[C/T]ACCAGAAGCAAATGCAGTGCGTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
YLPM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
YLPM1 - YLP motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019589.2 | 370 | Missense Mutation | CAC,TAC | H,Y 68 | NP_062535.2 | |
XM_005267860.3 | 370 | Missense Mutation | CAC,TAC | H,Y 68 | XP_005267917.1 | |
XM_011536966.2 | 370 | Intron | XP_011535268.1 | |||
XM_011536967.1 | 370 | Missense Mutation | CAC,TAC | H,Y 68 | XP_011535269.1 |