Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTACCACCAGGGCTGGAAGGAGA[C/T]CTCGAGAACCTTTGCAGGTAACGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604807 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLRT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLRT2 - fibronectin leucine rich transmembrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013231.4 | 655 | Intron | NP_037363.1 | |||
XM_005267490.3 | 655 | Intron | XP_005267547.1 | |||
XM_011536611.2 | 655 | Intron | XP_011534913.1 | |||
XM_017021129.1 | 655 | UTR 5 | XP_016876618.1 | |||
XM_017021130.1 | 655 | Intron | XP_016876619.1 | |||
XM_017021131.1 | 655 | Intron | XP_016876620.1 | |||
XM_017021132.1 | 655 | Intron | XP_016876621.1 | |||
XM_017021133.1 | 655 | Intron | XP_016876622.1 | |||
XM_017021134.1 | 655 | Intron | XP_016876623.1 | |||
XM_017021135.1 | 655 | Intron | XP_016876624.1 |
LOC100506731 - uncharacterized LOC100506731 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |