Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAATTTCCTACTCTGTGAAGTA[C/T]GTGTTAGAGGTGCAGAGATGAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611121 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLMN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLMN - calmin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024734.3 | 11330 | UTR 3 | NP_079010.2 | |||
XM_011537158.1 | 11330 | UTR 3 | XP_011535460.1 | |||
XM_011537159.2 | 11330 | Intron | XP_011535461.1 | |||
XM_017021646.1 | 11330 | UTR 3 | XP_016877135.1 | |||
XM_017021647.1 | 11330 | Intron | XP_016877136.1 |
DICER1-AS1 - DICER1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |