Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGCTCTCGTGGTGGCGGAAGTCA[C/T]CGTCGGCCGCCTTGAGGCCCTCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606558 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL11B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL11B - B-cell CLL/lymphoma 11B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282237.1 | 1826 | Missense Mutation | GAT,GGT | D,G 514 | NP_001269166.1 | |
NM_001282238.1 | 1826 | Missense Mutation | GAT,GGT | D,G 443 | NP_001269167.1 | |
NM_022898.2 | 1826 | Missense Mutation | GAT,GGT | D,G 444 | NP_075049.1 | |
NM_138576.3 | 1826 | Missense Mutation | GAT,GGT | D,G 515 | NP_612808.1 |