Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGCTTGCCCTGGCGAGTGGCCAG[C/T]GGGGCCCGCCGTTTTCGCCGCTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601146 | ||||||||||||||||||||
Literature Links: |
GDF5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GDF5 - growth differentiation factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000557.4 | 1411 | Silent Mutation | CCA,CCG | P,P 383 | NP_000548.2 | |
NM_001319138.1 | 1411 | Silent Mutation | CCA,CCG | P,P 383 | NP_001306067.1 | |
XM_011529075.2 | 1411 | Silent Mutation | CCA,CCG | P,P 383 | XP_011527377.1 |