Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAGTTCCATTGACTCTATTAA[C/T]ACATTTAAAAAATTGCAAAAATGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRCH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRCH2 - leucine rich repeats and calponin homology domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243963.1 | 2970 | UTR 3 | NP_001230892.1 | |||
NM_020871.3 | 2970 | UTR 3 | NP_065922.3 | |||
XM_006724724.3 | 2970 | UTR 3 | XP_006724787.2 | |||
XM_017029696.1 | 2970 | Intron | XP_016885185.1 | |||
XM_017029697.1 | 2970 | Intron | XP_016885186.1 |