Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTATGTTCACAGGCACCTGCTGC[C/T]GCTGCTGCTGCCTCTGCTCTTGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615741 MIM: 605622 MIM: 605675 | ||||||||||||||||||||
Literature Links: |
KIAA0141 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA0141 - KIAA0141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142603.2 | 4746 | Intron | NP_001136075.1 | |||
NM_014773.4 | 4746 | Intron | NP_055588.3 |
PCDH12 - protocadherin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016580.3 | 4746 | Missense Mutation | AGC,GGC | S,G 1179 | NP_057664.1 |
RNF14 - ring finger protein 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |