Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCGGTCCCATCTATCCACCTGGC[G/T]CCACAGTGCTGGTGGAAGGAGGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607431 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAZAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAZAP2 - DAZ associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136264.1 | 392 | Missense Mutation | GCC,TCC | A,S 80 | NP_001129736.1 | |
NM_001136266.1 | 392 | Intron | NP_001129738.1 | |||
NM_001136267.1 | 392 | Missense Mutation | GCC,TCC | A,S 70 | NP_001129739.1 | |
NM_001136268.1 | 392 | Intron | NP_001129740.1 | |||
NM_001136269.1 | 392 | Missense Mutation | GCC,TCC | A,S 102 | NP_001129741.1 | |
NM_014764.3 | 392 | Missense Mutation | GCC,TCC | A,S 102 | NP_055579.1 |
SMAGP - small cell adhesion glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |