Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAATCAGATGTTCCAGCTGGCTT[A/G]GAAGCAAGCAGACGTCCATGTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601300 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACVR1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACVR1B - activin A receptor type 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004302.4 | Intron | NP_004293.1 | ||||
NM_020327.3 | Intron | NP_064732.3 | ||||
NM_020328.3 | Intron | NP_064733.3 | ||||
XM_011538966.2 | Intron | XP_011537268.1 | ||||
XM_011538967.2 | Intron | XP_011537269.1 | ||||
XM_017020199.1 | Intron | XP_016875688.1 | ||||
XM_017020200.1 | Intron | XP_016875689.1 | ||||
XM_017020201.1 | Intron | XP_016875690.1 |