Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGCAAATTTGAGGATCAATTTT[A/T]AAAAAGTTATTACCTTTCTAAATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146680 | ||||||||||||||||||||
Literature Links: |
IDE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IDE - insulin degrading enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165946.1 | 2138 | UTR 3 | NP_001159418.1 | |||
NM_001322793.1 | 2138 | UTR 3 | NP_001309722.1 | |||
NM_001322794.1 | 2138 | UTR 3 | NP_001309723.1 | |||
NM_001322795.1 | 2138 | UTR 3 | NP_001309724.1 | |||
NM_001322796.1 | 2138 | UTR 3 | NP_001309725.1 | |||
NM_001322797.1 | 2138 | UTR 3 | NP_001309726.1 | |||
NM_004969.3 | 2138 | UTR 3 | NP_004960.2 | |||
XM_017016187.1 | 2138 | UTR 3 | XP_016871676.1 | |||
XM_017016188.1 | 2138 | UTR 3 | XP_016871677.1 | |||
XM_017016189.1 | 2138 | UTR 3 | XP_016871678.1 | |||
XM_017016190.1 | 2138 | UTR 3 | XP_016871679.1 |