Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGTGCGCCACGCTGGCCCGGAC[C/T]TTCGAGCAGCTCGTGGAGAACCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608428 MIM: 609672 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP26C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP26C1 - cytochrome P450 family 26 subfamily C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183374.2 | 639 | Silent Mutation | ACC,ACT | T,T 213 | NP_899230.2 |
EXOC6 - exocyst complex component 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |