Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGGAGGTTGTCAATGCAATTAG[A/G]TTACGGAATCTGCTTTATGGACTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610633 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MICU3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MICU3 - mitochondrial calcium uptake family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181723.2 | Intron | NP_859074.1 | ||||
XM_005273477.2 | Intron | XP_005273534.1 | ||||
XM_006716328.2 | Intron | XP_006716391.1 | ||||
XM_006716329.2 | Intron | XP_006716392.1 | ||||
XM_006716330.2 | Intron | XP_006716393.1 | ||||
XM_006716333.3 | Intron | XP_006716396.1 | ||||
XM_011544507.2 | Intron | XP_011542809.1 | ||||
XM_011544508.2 | Intron | XP_011542810.1 | ||||
XM_017013336.1 | Intron | XP_016868825.1 | ||||
XM_017013337.1 | Intron | XP_016868826.1 | ||||
XM_017013338.1 | Intron | XP_016868827.1 | ||||
XM_017013339.1 | Intron | XP_016868828.1 | ||||
XM_017013340.1 | Intron | XP_016868829.1 | ||||
XM_017013341.1 | Intron | XP_016868830.1 | ||||
XM_017013342.1 | Intron | XP_016868831.1 | ||||
XM_017013343.1 | Intron | XP_016868832.1 |