Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGCACACCTCATCGATGAGCAC[A/G]GCGCGCTCCGTACGTGAGGCGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607204 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PUM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PUM1 - pumilio RNA binding family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001020658.1 | 3404 | Silent Mutation | GCC,GCT | A,A 1097 | NP_001018494.1 | |
NM_014676.2 | 3404 | Silent Mutation | GCC,GCT | A,A 1095 | NP_055491.1 |
SNORD103A - small nucleolar RNA, C/D box 103A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |