Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCAAAAGGGATACACCTCCCTT[A/G]TTTGAGGAAGTGCTCCACCAGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609602 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLF17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLF17 - Kruppel like factor 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173484.3 | Intron | NP_775755.3 | ||||
XM_005270465.3 | Intron | XP_005270522.2 | ||||
XM_011540700.1 | Intron | XP_011539002.1 | ||||
XM_011540704.2 | Intron | XP_011539006.1 |