Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCCCAGGGAAGACTGCTTCTTG[C/T]GTAACGCCGGCCACAGAAAGAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF220 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF220 - ring finger protein 220 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319956.1 | 628 | UTR 5 | NP_001306885.1 | |||
NM_001319957.1 | 628 | Intron | NP_001306886.1 | |||
NM_018150.3 | 628 | UTR 5 | NP_060620.2 | |||
XM_005270992.1 | 628 | UTR 5 | XP_005271049.1 | |||
XM_005270993.1 | 628 | UTR 5 | XP_005271050.1 | |||
XM_005270996.1 | 628 | Intron | XP_005271053.1 | |||
XM_006710735.3 | 628 | UTR 5 | XP_006710798.1 | |||
XM_011541698.1 | 628 | UTR 5 | XP_011540000.1 | |||
XM_011541699.1 | 628 | UTR 5 | XP_011540001.1 | |||
XM_011541700.1 | 628 | UTR 5 | XP_011540002.1 | |||
XM_011541702.1 | 628 | Intron | XP_011540004.1 | |||
XM_017001623.1 | 628 | UTR 5 | XP_016857112.1 | |||
XM_017001624.1 | 628 | UTR 5 | XP_016857113.1 | |||
XM_017001625.1 | 628 | UTR 5 | XP_016857114.1 | |||
XM_017001626.1 | 628 | Intron | XP_016857115.1 |