Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTTGAATAGGTTAGCTAAGAGC[C/T]ATGACCACCACGCTGCCTTGCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604933 MIM: 604746 MIM: 613931 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUTYH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MUTYH - mutY DNA glycosylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TESK2 - testis-specific kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320800.1 | 3018 | UTR 3 | NP_001307729.1 | |||
NM_007170.2 | 3018 | UTR 3 | NP_009101.2 | |||
XM_005270355.3 | 3018 | UTR 3 | XP_005270412.1 | |||
XM_006710287.2 | 3018 | UTR 3 | XP_006710350.1 | |||
XM_011540497.1 | 3018 | UTR 3 | XP_011538799.1 | |||
XM_011540500.2 | 3018 | Intron | XP_011538802.1 | |||
XM_017000047.1 | 3018 | UTR 3 | XP_016855536.1 | |||
XM_017000048.1 | 3018 | UTR 3 | XP_016855537.1 | |||
XM_017000049.1 | 3018 | UTR 3 | XP_016855538.1 |
TOE1 - target of EGR1, member 1 (nuclear) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025077.3 | 3018 | Intron | NP_079353.3 | |||
XM_005270412.3 | 3018 | Intron | XP_005270469.1 | |||
XM_005270413.4 | 3018 | Intron | XP_005270470.1 | |||
XM_011540569.2 | 3018 | Intron | XP_011538871.1 |