Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCTCATTGTATGTAGCCCACTG[C/T]TGTATCATCTGCTAGGAGCAGTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 120960 | ||||||||||||||||||||
Literature Links: |
C8B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C8B - complement component 8, beta polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000066.3 | 1980 | UTR 3 | NP_000057.2 | |||
NM_001278543.1 | 1980 | UTR 3 | NP_001265472.1 | |||
NM_001278544.1 | 1980 | UTR 3 | NP_001265473.1 | |||
XM_017002235.1 | 1980 | UTR 3 | XP_016857724.1 |