Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCTGGCTTAAGGATTAGAAGCA[C/T]TGGCTTTCTAGACTATGGACAAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AK2 - adenylate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199199.1 | 2068 | UTR 3 | NP_001186128.1 | |||
NM_001319139.1 | 2068 | UTR 3 | NP_001306068.1 | |||
NM_001319140.1 | 2068 | Intron | NP_001306069.1 | |||
NM_001319141.1 | 2068 | UTR 3 | NP_001306070.1 | |||
NM_001319142.1 | 2068 | UTR 3 | NP_001306071.1 | |||
NM_001319143.1 | 2068 | Intron | NP_001306072.1 | |||
NM_001625.3 | 2068 | Intron | NP_001616.1 | |||
NM_013411.4 | 2068 | UTR 3 | NP_037543.1 |