Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAATAGGAATTAAAAGCCATTTA[A/G]TAAGTCCACATTCATTTCCTATTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 180069 | ||||||||||||||||||||
Literature Links: |
RPE65 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPE65 - RPE65, retinoid isomerohydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000329.2 | 2151 | UTR 3 | NP_000320.1 | |||
XM_017002027.1 | 2151 | UTR 3 | XP_016857516.1 |