Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAACATTGTACAATATAAGCGGCA[A/G]TAAGTTACTGATATCTGCTGACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615146 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP33 - ubiquitin specific peptidase 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015017.4 | 4132 | UTR 3 | NP_055832.3 | |||
NM_201624.2 | 4132 | UTR 3 | NP_963918.1 | |||
NM_201626.2 | 4132 | Intron | NP_963920.1 | |||
XM_005270648.1 | 4132 | UTR 3 | XP_005270705.1 | |||
XM_005270649.1 | 4132 | UTR 3 | XP_005270706.1 | |||
XM_011541055.1 | 4132 | UTR 3 | XP_011539357.1 | |||
XM_011541056.2 | 4132 | Intron | XP_011539358.1 | |||
XM_017000722.1 | 4132 | UTR 3 | XP_016856211.1 | |||
XM_017000723.1 | 4132 | UTR 3 | XP_016856212.1 | |||
XM_017000724.1 | 4132 | UTR 3 | XP_016856213.1 | |||
XM_017000725.1 | 4132 | UTR 3 | XP_016856214.1 | |||
XM_017000726.1 | 4132 | UTR 3 | XP_016856215.1 | |||
XM_017000727.1 | 4132 | UTR 3 | XP_016856216.1 | |||
XM_017000728.1 | 4132 | UTR 3 | XP_016856217.1 | |||
XM_017000729.1 | 4132 | UTR 3 | XP_016856218.1 |