Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTCTCCTCGGGTAGCTCATAGT[A/C]CTCCGCCTCACGCTCCATCTGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611413 | ||||||||||||||||||||
Literature Links: |
DLGAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLGAP3 - DLG associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080418.2 | 2442 | Missense Mutation | GAC,TAC | D,Y 822 | NP_001073887.1 | |
XM_011541879.2 | 2442 | Missense Mutation | GAC,TAC | D,Y 822 | XP_011540181.1 | |
XM_011541880.2 | 2442 | Missense Mutation | GAC,TAC | D,Y 325 | XP_011540182.1 |
SMIM12 - small integral membrane protein 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |