Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGATTGGTTCATTTTAAAAGTCA[C/T]TTTCTCTGTGTTTCGTTTTTGATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAMD13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAMD13 - sterile alpha motif domain containing 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010971.2 | Intron | NP_001010971.1 | ||||
NM_001134663.1 | Intron | NP_001128135.1 | ||||
NM_001134664.1 | Intron | NP_001128136.1 | ||||
XM_017000377.1 | Intron | XP_016855866.1 |