Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCATCCAGGTAGTATTCATCATC[C/T]GTGGTTGTGTCTGTGTCACAATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612878 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXPH5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXPH5 - exophilin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144763.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1898 | NP_001138235.1 | |
NM_001144764.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1818 | NP_001138236.1 | |
NM_001144765.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1786 | NP_001138237.1 | |
NM_001308019.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1967 | NP_001294948.1 | |
NM_015065.2 | 6033 | Silent Mutation | ACA,ACG | T,T 1974 | NP_055880.2 | |
XM_011542696.2 | 6033 | Silent Mutation | ACA,ACG | T,T 1898 | XP_011540998.1 | |
XM_011542698.2 | 6033 | Silent Mutation | ACA,ACG | T,T 1898 | XP_011541000.1 | |
XM_011542700.2 | 6033 | Silent Mutation | ACA,ACG | T,T 1898 | XP_011541002.1 | |
XM_017017397.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1898 | XP_016872886.1 | |
XM_017017398.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1897 | XP_016872887.1 | |
XM_017017399.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1898 | XP_016872888.1 | |
XM_017017400.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1898 | XP_016872889.1 | |
XM_017017401.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1897 | XP_016872890.1 | |
XM_017017402.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1818 | XP_016872891.1 | |
XM_017017403.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1817 | XP_016872892.1 | |
XM_017017404.1 | 6033 | Silent Mutation | ACA,ACG | T,T 1786 | XP_016872893.1 |