Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCATTTCCTAATTAGTCACCAA[C/T]ATTGATCCAGATCCTCGTCCCTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602690 MIM: 606659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf57 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf57 - chromosome 11 open reading frame 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082969.1 | Intron | NP_001076438.1 | ||||
NM_001082970.1 | Intron | NP_001076439.1 | ||||
NM_001301017.1 | Intron | NP_001287946.1 | ||||
NM_001301019.1 | Intron | NP_001287948.1 | ||||
NM_001301021.1 | Intron | NP_001287950.1 | ||||
NM_018195.3 | Intron | NP_060665.3 |
SDHD - succinate dehydrogenase complex subunit D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIMM8B - translocase of inner mitochondrial membrane 8 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012459.2 | Intron | NP_036591.2 |