Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTTTGCATAAGGAATATAACCT[A/G]TCACCATCTTCAGTAGATATGTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603954 | ||||||||||||||||||||
Literature Links: |
ZW10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZW10 - zw10 kinetochore protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004724.3 | 1885 | Silent Mutation | GAC,GAT | D,D 680 | NP_004715.1 | |
XM_017018558.1 | 1885 | Silent Mutation | GAC,GAT | D,D 616 | XP_016874047.1 | |
XM_017018559.1 | 1885 | Silent Mutation | GAC,GAT | D,D 460 | XP_016874048.1 |