Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGGCAGCCTCTGATGGGGCAAC[A/G]GCTGAGGGTGGGGGCAGTGTGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610658 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM29 - tripartite motif containing 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012101.3 | 2123 | UTR 3 | NP_036233.2 | |||
XM_005271490.1 | 2123 | UTR 3 | XP_005271547.1 | |||
XM_011542729.1 | 2123 | UTR 3 | XP_011541031.1 | |||
XM_011542732.1 | 2123 | UTR 3 | XP_011541034.1 | |||
XM_011542733.1 | 2123 | UTR 3 | XP_011541035.1 | |||
XM_017017453.1 | 2123 | UTR 3 | XP_016872942.1 | |||
XM_017017454.1 | 2123 | UTR 3 | XP_016872943.1 |