Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAATAACCTTTCAAATTTGAGTG[A/C]CTCTCCTTTGCAAACAACTGCAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC37A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC37A2 - solute carrier family 37 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM218 - transmembrane protein 218 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080546.2 | 1399 | UTR 3 | NP_001074015.1 | |||
NM_001258238.1 | 1399 | UTR 3 | NP_001245167.1 | |||
NM_001258239.1 | 1399 | UTR 3 | NP_001245168.1 | |||
NM_001258240.1 | 1399 | UTR 3 | NP_001245169.1 | |||
NM_001258241.1 | 1399 | UTR 3 | NP_001245170.1 | |||
NM_001258242.1 | 1399 | UTR 3 | NP_001245171.1 | |||
NM_001258243.1 | 1399 | UTR 3 | NP_001245172.1 | |||
NM_001258244.1 | 1399 | UTR 3 | NP_001245173.1 | |||
NM_001258245.1 | 1399 | UTR 3 | NP_001245174.1 | |||
NM_001258246.1 | 1399 | UTR 3 | NP_001245175.1 | |||
NM_001258247.1 | 1399 | UTR 3 | NP_001245176.1 | |||
XM_006718779.3 | 1399 | UTR 3 | XP_006718842.1 | |||
XM_006718780.3 | 1399 | UTR 3 | XP_006718843.1 | |||
XM_006718781.3 | 1399 | UTR 3 | XP_006718844.1 | |||
XM_006718783.3 | 1399 | UTR 3 | XP_006718846.1 | |||
XM_017017322.1 | 1399 | UTR 3 | XP_016872811.1 | |||
XM_017017323.1 | 1399 | UTR 3 | XP_016872812.1 | |||
XM_017017324.1 | 1399 | UTR 3 | XP_016872813.1 |