Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTTTGGCTCTCCCCGCCTGTT[C/T]TTTGCACATTAAAAATGAAAAAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603078 MIM: 601134 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHEK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHEK1 - checkpoint kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114121.2 | 274 | UTR 5 | NP_001107593.1 | |||
NM_001114122.2 | 274 | UTR 5 | NP_001107594.1 | |||
NM_001244846.1 | 274 | UTR 5 | NP_001231775.1 | |||
NM_001274.5 | 274 | Intron | NP_001265.2 | |||
XM_011542560.2 | 274 | UTR 5 | XP_011540862.1 | |||
XM_011542561.2 | 274 | UTR 5 | XP_011540863.1 | |||
XM_011542562.2 | 274 | Intron | XP_011540864.1 | |||
XM_011542563.2 | 274 | Intron | XP_011540865.1 | |||
XM_017017146.1 | 274 | Intron | XP_016872635.1 |
STT3A - STT3A, catalytic subunit of the oligosaccharyltransferase complex | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |