Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCAACTCTAAGCTCGGAGAAGA[C/T]GGGAACACTGGTAATCTCCACAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607311 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGR - progesterone receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000926.4 | 9831 | UTR 3 | NP_000917.3 | |||
NM_001202474.3 | 9831 | UTR 3 | NP_001189403.1 | |||
NM_001271161.2 | 9831 | UTR 3 | NP_001258090.1 | |||
NM_001271162.1 | 9831 | UTR 3 | NP_001258091.1 | |||
XM_006718858.3 | 9831 | Intron | XP_006718921.1 | |||
XM_011542869.2 | 9831 | Intron | XP_011541171.1 |