Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCAAACCCAGGATCTCCGCCGC[C/G]AGATCCACCCCGGCGCCCCCGAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102574 | ||||||||||||||||||||
Literature Links: |
ACTN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTN3 - actinin alpha 3 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104.3 | 332 | Intron | NP_001095.2 | |||
NM_001258371.2 | 332 | Silent Mutation | GCC,GCG | A,A 72 | NP_001245300.2 |
ZDHHC24 - zinc finger DHHC-type containing 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207340.1 | 332 | Intron | NP_997223.1 | |||
XM_005273874.3 | 332 | Intron | XP_005273931.1 | |||
XM_011544893.2 | 332 | Intron | XP_011543195.1 | |||
XM_011544894.1 | 332 | Intron | XP_011543196.1 |