Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGAGCAGAAAGGTCCCAAACTCA[C/T]GAAGAGGAACCTGATTTTTCAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605657 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM2A - lysine demethylase 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256405.1 | Intron | NP_001243334.1 | ||||
NM_012308.2 | Intron | NP_036440.1 | ||||
XM_006718480.3 | Intron | XP_006718543.1 | ||||
XM_011544860.2 | Intron | XP_011543162.1 | ||||
XM_011544861.1 | Intron | XP_011543163.1 | ||||
XM_017017391.1 | Intron | XP_016872880.1 | ||||
XM_017017392.1 | Intron | XP_016872881.1 |
LOC107984341 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |