Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTCATGGAAAGGAGTCGATGGG[A/G]CAAGAAGAGAAGTATTTGAACAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600843 MIM: 604328 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
P2RX3 - purinergic receptor P2X 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002559.3 | 168 | Intron | NP_002550.2 | |||
XM_011545069.2 | 168 | UTR 5 | XP_011543371.1 | |||
XM_011545070.2 | 168 | Intron | XP_011543372.1 | |||
XM_011545071.2 | 168 | Intron | XP_011543373.1 | |||
XM_011545072.2 | 168 | Intron | XP_011543374.1 | |||
XM_011545073.1 | 168 | Intron | XP_011543375.1 |
SSRP1 - structure specific recognition protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003146.2 | 168 | Intron | NP_003137.1 | |||
XM_017018180.1 | 168 | Intron | XP_016873669.1 | |||
XM_017018181.1 | 168 | Intron | XP_016873670.1 |