Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTAGTGGACAGAGATGGCCACC[A/G]ATTGCAGTGGGGATATTTCATGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHNAK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHNAK - AHNAK nucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001620.2 | Intron | NP_001611.1 | ||||
NM_024060.3 | Intron | NP_076965.2 | ||||
XM_005274240.1 | Intron | XP_005274297.1 | ||||
XM_005274241.2 | Intron | XP_005274298.1 | ||||
XM_005274245.1 | Intron | XP_005274302.1 | ||||
XM_017018270.1 | Intron | XP_016873759.1 | ||||
XM_017018271.1 | Intron | XP_016873760.1 | ||||
XM_017018272.1 | Intron | XP_016873761.1 | ||||
XM_017018273.1 | Intron | XP_016873762.1 |