Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAAATAGGAGTTTCTCGTCCTC[C/T]AGGAGGACCTGGTTTTCTAGTTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF485 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF485 - zinc finger protein 485 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318140.1 | Intron | NP_001305069.1 | ||||
NM_001318141.1 | Intron | NP_001305070.1 | ||||
NM_001318142.1 | Intron | NP_001305071.1 | ||||
NM_001318143.1 | Intron | NP_001305072.1 | ||||
NM_145312.3 | Intron | NP_660355.2 | ||||
XM_011539498.2 | Intron | XP_011537800.1 |