Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAGCTGAGCCCATTCCTTTGGTG[G/T]ATGCGGAAGACGCTGTCATCGTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612570 | ||||||||||||||||||||
Literature Links: |
FBN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBN2 - fibrillin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001999.3 | 9143 | Silent Mutation | ATA,ATC | I,I 2847 | NP_001990.2 | |
XM_017009228.1 | 9143 | Silent Mutation | ATA,ATC | I,I 2796 | XP_016864717.1 |