Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGGTGGATTAGTTCCTCCTGA[A/G]ACTCAATCACGTCCCGCAGTTGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DNAJC18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC18 - DnaJ heat shock protein family (Hsp40) member C18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PROB1 - proline rich basic protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA24 - spermatogenesis associated 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194296.1 | 110 | Missense Mutation | TCT,TTT | S,F 28 | NP_919272.1 | |
XM_005271916.4 | 110 | Missense Mutation | TCT,TTT | S,F 28 | XP_005271973.1 | |
XM_011543252.2 | 110 | UTR 5 | XP_011541554.1 | |||
XM_011543253.2 | 110 | UTR 5 | XP_011541555.1 | |||
XM_011543254.2 | 110 | Missense Mutation | TCT,TTT | S,F 28 | XP_011541556.1 |