Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATAAACTGTAGCATATTCACAAA[C/T]TGAAATAGTACTTAGTACTCAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613365 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO6A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLCO6A1 - solute carrier organic anion transporter family member 6A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289002.1 | Intron | NP_001275931.1 | ||||
NM_001289004.1 | Intron | NP_001275933.1 | ||||
NM_001308014.1 | Intron | NP_001294943.1 | ||||
NM_173488.4 | Intron | NP_775759.3 | ||||
XM_005271874.3 | Intron | XP_005271931.1 | ||||
XM_011543147.2 | Intron | XP_011541449.1 | ||||
XM_011543148.2 | Intron | XP_011541450.1 | ||||
XM_011543150.1 | Intron | XP_011541452.1 | ||||
XM_011543153.2 | Intron | XP_011541455.1 | ||||
XM_011543154.2 | Intron | XP_011541456.1 |