Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCCTCTCATGACATTCCTGGC[A/G]TGCATGTCACAAGTTGCTAGGATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615365 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AK8 - adenylate kinase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPACA9 - sperm acrosome associated 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316897.1 | Intron | NP_001303826.1 | ||||
NM_001316898.1 | Intron | NP_001303827.1 | ||||
NM_001316900.1 | Intron | NP_001303829.1 | ||||
NM_018956.4 | Intron | NP_061829.3 | ||||
XM_017014231.1 | Intron | XP_016869720.1 |