Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCAGCCTCAGCCTCCTCGGGCAC[A/C]CGCACAGGCACCGCGCCACGGTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605247 MIM: 180530 MIM: 609302 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PANO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PANO1 - proapoptotic nucleolar protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIDD1 - p53-induced death domain protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145886.3 | 3350 | Silent Mutation | CGG,CGT | R,R 731 | NP_665893.2 | |
NM_145887.3 | 3350 | Silent Mutation | CGG,CGT | R,R 714 | NP_665894.2 | |
XM_005253005.4 | 3350 | Silent Mutation | CGG,CGT | R,R 755 | XP_005253062.1 | |
XM_005253006.4 | 3350 | Silent Mutation | CGG,CGT | R,R 755 | XP_005253063.1 | |
XM_005253007.3 | 3350 | Silent Mutation | CGG,CGT | R,R 755 | XP_005253064.1 | |
XM_005253008.4 | 3350 | Silent Mutation | CGG,CGT | R,R 609 | XP_005253065.1 | |
XM_011520209.2 | 3350 | Silent Mutation | CGG,CGT | R,R 741 | XP_011518511.1 | |
XM_011520210.2 | 3350 | Silent Mutation | CGG,CGT | R,R 731 | XP_011518512.1 | |
XM_011520211.2 | 3350 | Silent Mutation | CGG,CGT | R,R 714 | XP_011518513.1 | |
XM_011520212.1 | 3350 | Silent Mutation | CGG,CGT | R,R 609 | XP_011518514.1 | |
XM_011520213.2 | 3350 | Silent Mutation | CGG,CGT | R,R 442 | XP_011518515.1 | |
XM_017017993.1 | 3350 | Silent Mutation | CGG,CGT | R,R 418 | XP_016873482.1 |
RPLP2 - ribosomal protein lateral stalk subunit P2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A22 - solute carrier family 25 member 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |