Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCGCAGAGTCGCAGGAGGCGGCG[A/C]CGTTATCAGGTAGGGCGCCCAGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608159 | ||||||||||||||||||||
Literature Links: |
PRSS21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRSS21 - protease, serine 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270452.1 | 108 | Missense Mutation | ACG,CCG | T,P 28 | NP_001257381.1 | |
NM_006799.3 | 108 | Missense Mutation | ACG,CCG | T,P 28 | NP_006790.1 | |
NM_144956.2 | 108 | Missense Mutation | ACG,CCG | T,P 28 | NP_659205.1 | |
NM_144957.2 | 108 | Missense Mutation | ACG,CCG | T,P 28 | NP_659206.1 | |
XM_017022875.1 | 108 | Missense Mutation | ACG,CCG | T,P 28 | XP_016878364.1 | |
XM_017022876.1 | 108 | Missense Mutation | ACG,CCG | T,P 28 | XP_016878365.1 | |
XM_017022877.1 | 108 | Missense Mutation | ACG,CCG | T,P 28 | XP_016878366.1 | |
XM_017022878.1 | 108 | Missense Mutation | ACG,CCG | T,P 28 | XP_016878367.1 |