Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCGGGTACACTGAGGATGGCTG[C/T]GCGTTCATGCTCCGACAGGCTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156565 MIM: 600928 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPG - N-methylpurine DNA glycosylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPRL3 - NPR3 like, GATOR1 complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039476.2 | 1810 | Missense Mutation | ACA,GCA | T,A 317 | NP_001034565.1 | |
NM_001077350.2 | 1810 | Missense Mutation | ACA,GCA | T,A 496 | NP_001070818.1 | |
NM_001243247.1 | 1810 | Missense Mutation | ACA,GCA | T,A 418 | NP_001230176.1 | |
NM_001243248.1 | 1810 | Missense Mutation | ACA,GCA | T,A 471 | NP_001230177.1 | |
NM_001243249.1 | 1810 | Missense Mutation | ACA,GCA | T,A 471 | NP_001230178.1 |