Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACTGGAAAGGAAATAGATTCTT[A/G]AAGAAATAAAAATAAGCACAATTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 115450 MIM: 115460 | ||||||||||||||||||||
Literature Links: |
CSN1S1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSN1S1 - casein alpha s1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CSN2 - casein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302770.1 | 1021 | UTR 3 | NP_001289699.1 | |||
NM_001891.3 | 1021 | UTR 3 | NP_001882.1 | |||
XM_017007760.1 | 1021 | UTR 3 | XP_016863249.1 |