Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCACGCGCTTGGCCTCGCCGATG[C/T]GGATACGCTTGATGGGGTCGGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGK223 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGK223 - homolog of rat pragma of Rnd2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080826.2 | 4625 | Missense Mutation | CAC,CGC | H,R 1317 | NP_001074295.2 | |
XM_005272369.4 | 4625 | Missense Mutation | CAC,CGC | H,R 1317 | XP_005272426.2 | |
XM_005272370.4 | 4625 | Missense Mutation | CAC,CGC | H,R 1317 | XP_005272427.2 | |
XM_011543813.2 | 4625 | Intron | XP_011542115.1 |