Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTGAAGAAATGGGCCCAATAAT[A/G]TGATTCCTAATCCTTCTGGATCTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611649 | ||||||||||||||||||||
Literature Links: |
FAM188A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM188A - family with sequence similarity 188 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318330.1 | 1287 | Missense Mutation | ACA,ATA | T,I 186 | NP_001305259.1 | |
NM_024948.3 | 1287 | Missense Mutation | ACA,ATA | T,I 359 | NP_079224.1 | |
XM_005252600.2 | 1287 | Missense Mutation | ACA,ATA | T,I 186 | XP_005252657.1 | |
XM_005252602.2 | 1287 | Missense Mutation | ACA,ATA | T,I 186 | XP_005252659.1 | |
XM_006717508.2 | 1287 | Missense Mutation | ACA,ATA | T,I 332 | XP_006717571.1 | |
XM_011519690.2 | 1287 | Missense Mutation | ACA,ATA | T,I 359 | XP_011517992.1 | |
XM_011519691.2 | 1287 | Missense Mutation | ACA,ATA | T,I 359 | XP_011517993.1 | |
XM_011519693.1 | 1287 | Missense Mutation | ACA,ATA | T,I 212 | XP_011517995.1 | |
XM_011519694.1 | 1287 | Missense Mutation | ACA,ATA | T,I 186 | XP_011517996.1 | |
XM_017016671.1 | 1287 | Missense Mutation | ACA,ATA | T,I 332 | XP_016872160.1 | |
XM_017016672.1 | 1287 | Missense Mutation | ACA,ATA | T,I 212 | XP_016872161.1 | |
XM_017016673.1 | 1287 | Missense Mutation | ACA,ATA | T,I 186 | XP_016872162.1 | |
XM_017016674.1 | 1287 | Missense Mutation | ACA,ATA | T,I 159 | XP_016872163.1 | |
XM_017016675.1 | 1287 | Missense Mutation | ACA,ATA | T,I 159 | XP_016872164.1 | |
XM_017016676.1 | 1287 | Missense Mutation | ACA,ATA | T,I 159 | XP_016872165.1 |