Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAGCAAAGGGAGACCTACCCCA[A/C]CCCAAAAAGGGAAAGTTCTTTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 200350 MIM: 602276 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACACA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACACA - acetyl-CoA carboxylase alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TADA2A - transcriptional adaptor 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166105.2 | Intron | NP_001159577.2 | ||||
NM_001291918.1 | Intron | NP_001278847.1 | ||||
NM_001488.4 | Intron | NP_001479.4 | ||||
NM_133439.3 | Intron | NP_597683.3 | ||||
XM_006722044.3 | Intron | XP_006722107.1 | ||||
XM_011525153.2 | Intron | XP_011523455.1 | ||||
XM_011525154.2 | Intron | XP_011523456.1 | ||||
XM_011525155.2 | Intron | XP_011523457.1 | ||||
XM_017024981.1 | Intron | XP_016880470.1 | ||||
XM_017024982.1 | Intron | XP_016880471.1 | ||||
XM_017024983.1 | Intron | XP_016880472.1 | ||||
XM_017024984.1 | Intron | XP_016880473.1 | ||||
XM_017024985.1 | Intron | XP_016880474.1 | ||||
XM_017024986.1 | Intron | XP_016880475.1 |