Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGATTACCTGAGGTCAGGAGTT[C/T]GAGACCAGCCTGCCCAACATGGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300573 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF674 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF674 - zinc finger protein 674 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039891.2 | 3134 | UTR 3 | NP_001034980.1 | |||
NM_001146291.1 | 3134 | UTR 3 | NP_001139763.1 | |||
NM_001190417.1 | 3134 | UTR 3 | NP_001177346.1 | |||
XM_011543941.2 | 3134 | Intron | XP_011542243.1 | |||
XM_011543943.2 | 3134 | Intron | XP_011542245.1 | |||
XM_011543944.2 | 3134 | Intron | XP_011542246.1 | |||
XM_011543945.2 | 3134 | Intron | XP_011542247.1 | |||
XM_017029728.1 | 3134 | Intron | XP_016885217.1 | |||
XM_017029729.1 | 3134 | Intron | XP_016885218.1 | |||
XM_017029730.1 | 3134 | Intron | XP_016885219.1 | |||
XM_017029731.1 | 3134 | Intron | XP_016885220.1 | |||
XM_017029732.1 | 3134 | Intron | XP_016885221.1 | |||
XM_017029733.1 | 3134 | Intron | XP_016885222.1 |