Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATTAGCAAAAGATAAAAGGCTG[C/T]AATCAAAAACAGAATAAGGGAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601924 MIM: 600279 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPA - coatomer protein complex subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098398.1 | 5377 | UTR 3 | NP_001091868.1 | |||
NM_004371.3 | 5377 | UTR 3 | NP_004362.2 |
PEX19 - peroxisomal biogenesis factor 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |