Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAACTTGAGTGAAGGGTCAAGAT[C/T]TGGATTTCACCTTTCCTCTGAGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606841 MIM: 604618 MIM: 610554 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEDD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEDD - death effector domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039711.1 | Intron | NP_001034800.1 | ||||
NM_001039712.1 | Intron | NP_001034801.1 | ||||
NM_032998.2 | Intron | NP_127491.1 | ||||
XM_005245597.3 | Intron | XP_005245654.1 | ||||
XM_005245598.4 | Intron | XP_005245655.1 | ||||
XM_005245599.2 | Intron | XP_005245656.1 | ||||
XM_005245600.2 | Intron | XP_005245657.1 | ||||
XM_017002802.1 | Intron | XP_016858291.1 |
NIT1 - nitrilase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFC1 - ubiquitin-fold modifier conjugating enzyme 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016406.3 | Intron | NP_057490.2 | ||||
XM_005245254.1 | Intron | XP_005245311.1 | ||||
XM_017001450.1 | Intron | XP_016856939.1 | ||||
XM_017001451.1 | Intron | XP_016856940.1 |