Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGGGCAGTGTGGGTGTGACAGA[A/G]ATCAACAAGACTGCAGGAGCACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf111 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf111 - chromosome 1 open reading frame 111 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1orf226 - chromosome 1 open reading frame 226 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085375.1 | 415 | Silent Mutation | GAA,GAG | E,E 81 | NP_001078844.1 | |
NM_001135240.1 | 415 | Silent Mutation | GAA,GAG | E,E 124 | NP_001128712.1 |