Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGCGGAGCCGCCGCGTTTTCTG[A/G]AGGCCTTCGGGCGGCTGTGGCAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608849 | ||||||||||||||||||||
Literature Links: |
UHMK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UHMK1 - U2AF homology motif (UHM) kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184763.1 | 245 | Intron | NP_001171692.1 | |||
NM_144624.2 | 245 | Missense Mutation | AAG,GAG | K,E 17 | NP_653225.2 | |
NM_175866.4 | 245 | Missense Mutation | AAG,GAG | K,E 17 | NP_787062.1 |